Ethyl-5-amino-1-methyl-1H-pyrazol-4-carboxylat

Lieferant: FLUOROCHEM

Synonyme: Ethyl 5-amino-1-methyl-1H-pyrazole-4-carboxylate

CFF617.1 005506-1G 005506-5G
CHMPCFF617.1EA 532 EUR
CHMPCFF617.1 FLUO005506-1G FLUO005506-5G
Ethyl-5-amino-1-methyl-1H-pyrazol-4-carboxylat
Ethyl-5-amino-1-methyl-1H-pyrazol-4-carboxylat
Formel: C₇H₁₁N₃O₂
MDL: MFCD00051652
CAS-Nummer: 31037-02-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR