4-Cyan-2-ethylanilin

Lieferant: FLUOROCHEM

CFF551.5 CFF551.25 CFF551.50 005508-1G 005508-5G
CHMPCFF551.5EA 401 EUR
CHMPCFF551.5 CHMPCFF551.25 CHMPCFF551.50 FLUO005508-1G FLUO005508-5G
4-Cyan-2-ethylanilin
4-Cyan-2-ethylanilin
Formel: C₉H₁₀N₂
MDL: MFCD00041431
CAS-Nummer: 170230-87-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR