4-Cyan-2-ethylanilin
Lieferant: FLUOROCHEM
CFF551.5
CFF551.25
CFF551.50
005508-1G
005508-5G
CHMPCFF551.5EA
401
EUR
CHMPCFF551.5
CHMPCFF551.25
CHMPCFF551.50
FLUO005508-1G
FLUO005508-5G
4-Cyan-2-ethylanilin
4-Cyan-2-ethylanilin
Formel:
C₉H₁₀N₂ |
MDL:
MFCD00041431 CAS-Nummer: 170230-87-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...