2,4,5-Trifluorbenzolsulfonamid

Lieferant: FLUOROCHEM

Synonyme: 2,4,5-Trifluorbenzolsulfonsäureamid

CFF1272. 005527-1G
CHMPCFF1272.EA 0 EUR
CHMPCFF1272. FLUO005527-1G
2,4,5-Trifluorbenzolsulfonamid
2,4,5-Trifluorbenzolsulfonamid
Formel: C₆H₄F₃NO₂S
Molecular Weight: 211,16 g/mol
MDL: MFCD01569462
CAS-Nummer: 287172-63-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR