4-(Trifluormethoxy)benzolsulfonamid
Lieferant: FLUOROCHEM
Synonyme:
4-(Trifluormethoxy)benzolsulfonsäureamid
CFF1279.
005531-1G
005531-5G
CHMPCFF1279.EA
0
EUR
CHMPCFF1279.
FLUO005531-1G
FLUO005531-5G
4-(Trifluormethoxy)benzolsulfonamid
4-(Trifluormethoxy)benzolsulfonamid
Formel:
C₇H₆F₃NO₃S Molecular Weight: 241.19 g/mol |
MDL:
MFCD00798381 CAS-Nummer: 1513-45-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...