1-Brom-2-chlor-4-fluorbenzol

Lieferant: FLUOROCHEM
Achtung

Synonyme: 4-Brom-3-chlor-1-fluorbenzol

005539-1G 005539-5G 005539-25G
FLUO005539-1GEA 31.1 EUR
FLUO005539-1G FLUO005539-5G FLUO005539-25G
1-Brom-2-chlor-4-fluorbenzol
1-Bromo-2-chloro-4-fluorobenzene
Formel: C₆H₃BrClF
Molecular Weight: 209.44 g/mol
MDL: MFCD00142587
CAS-Nummer: 110407-59-5

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR