2,7-Dimethylnaphthalin
Lieferant: FLUOROCHEM
005541-1G
FLUO005541-1GEA
326
EUR
FLUO005541-1G
2,7-Dimethylnaphthalin
2,7-Dimethylnaphthalene
Formel:
C₁₂H₁₂ Molecular Weight: 156,23 g/mol Siedepunkt: 263 °C (1013 hPa) Schmelzpunkt: 94…95 °C Dichte: 1,003 g/cm³ (20 °C) |
MDL:
MFCD00004121 CAS-Nummer: 582-16-1 EINECS: 209-477-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...