trans,trans-Dibenzylidenaceton

Lieferant: FLUOROCHEM

005548-25G 005548-100G
FLUO005548-25GEA 79.1 EUR
FLUO005548-25G FLUO005548-100G
trans,trans-Dibenzylidenaceton
trans,trans-Dibenzylidenaceton
Formel: C₁₇H₁₄O
MDL: MFCD00004790
CAS-Nummer: 35225-79-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR