5-(Trifluormethoxy)salicylaldehyd

Lieferant: FLUOROCHEM

CFF1281. 005558-1G 005558-5G
CHMPCFF1281.EA 0 EUR
CHMPCFF1281. FLUO005558-1G FLUO005558-5G
5-(Trifluormethoxy)salicylaldehyd
5-(Trifluormethoxy)salicylaldehyd
Formel: C₈H₅F₃O₃
MDL: MFCD00075249
CAS-Nummer: 93249-62-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR