2,5-Diiodbenzoesäure

Lieferant: FLUOROCHEM

005559-5G
FLUO005559-5GEA 57.9 EUR
FLUO005559-5G
2,5-Diiodbenzoesäure
2,5-Diiodbenzoesäure
Formel: C₇H₄I₂O₂
Molecular Weight: 373,92 g/mol
MDL: MFCD00045801
CAS-Nummer: 14192-12-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR