3,5-Dichlorbenzoesäure
Lieferant: FLUOROCHEM
|
Achtung
|
CFF1007.
005560-25G
005560-100G
CHMPCFF1007.EA
0
EUR
CHMPCFF1007.
FLUO005560-25G
FLUO005560-100G
3,5-Dichlorbenzoesäure
3,5-Dichlorobenzoic acid
Formel:
C₇H₄Cl₂O₂ Molecular Weight: 191.01 g/mol Siedepunkt: 309 Schmelzpunkt: 183…185 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002494 CAS-Nummer: 51-36-5 EINECS: 200-092-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...