5-Amino-2-(trifluormethyl)benzimidazol

Lieferant: FLUOROCHEM

005565.5G 005565-1G
FLUO005565.5GEA 2010 EUR
FLUO005565.5G FLUO005565-1G
5-Amino-2-(trifluormethyl)benzimidazol
5-Amino-2-(trifluoromethyl)benzimidazole
Formel: C₈H₆F₃N₃
Molecular Weight: 201,15 g/mol
MDL: MFCD01632182
CAS-Nummer: 3671-66-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR