1-Chlor-2,4-bis(trifluormethyl)benzol

Lieferant: FLUOROCHEM

005570-5G 005570-1G
FLUO005570-5GEA 183 EUR
FLUO005570-5G FLUO005570-1G
1-Chlor-2,4-bis(trifluormethyl)benzol
1-Chlor-2,4-bis(trifluormethyl)benzol
Formel: C₈H₃ClF₆
MDL: MFCD01631638
CAS-Nummer: 327-76-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR