3,5-Bis(trifluormethyl)-o-phenylendiamin

Lieferant: FLUOROCHEM

Synonyme: 3,5-Bis(trifluoromethyl)-1,2-diaminobenzene

005571-1G
FLUO005571-1GEA 51.8 EUR
FLUO005571-1G
3,5-Bis(trifluormethyl)-o-phenylendiamin
3,5-Bis(trifluormethyl)-o-phenylendiamin
Formel: C₈H₆F₆N₂
MDL: MFCD01631430
CAS-Nummer: 367-65-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR