5-Brom-2,3-difluorphenol

Lieferant: FLUOROCHEM
Achtung

005581-5G 005581-25G
FLUO005581-5GEA 36.9 EUR
FLUO005581-5G FLUO005581-25G
5-Brom-2,3-difluorphenol
5-Brom-2,3-difluorphenol
Formel: C₆H₃BrF₂O
MDL: MFCD01631350
CAS-Nummer: 186590-26-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR