3-Chlor-2,4-difluorbenzoesäure

Lieferant: FLUOROCHEM
Achtung

005591-5G 005591-10G 005591-1G
FLUO005591-5GEA 48.8 EUR
FLUO005591-5G FLUO005591-10G FLUO005591-1G
3-Chlor-2,4-difluorbenzoesäure
3-Chloro-2,4-difluorobenzoic acid
Formel: C₇H₃ClF₂O₂
Molecular Weight: 192.55 g/mol
MDL: MFCD01631387
CAS-Nummer: 154257-75-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR