(2-Chlor-3,6-difluorphenyl)essigsäure

Lieferant: FLUOROCHEM

005606-1G 005606-5G
FLUO005606-1GEA 209 EUR
FLUO005606-1G FLUO005606-5G
(2-Chlor-3,6-difluorphenyl)essigsäure
(2-Chlor-3,6-difluorphenyl)essigsäure
Formel: C₈H₅ClF₂O₂
Molecular Weight: 206.58 g/mol
MDL: MFCD01631448
CAS-Nummer: 261762-52-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR