3-Chlor-2-fluorbenzaldehyd

Lieferant: FLUOROCHEM
Achtung

005613-5G 005613-10G 005613-25G
FLUO005613-5GEA 31.1 EUR
FLUO005613-5G FLUO005613-10G FLUO005613-25G
3-Chlor-2-fluorbenzaldehyd
3-Chloro-2-fluorobenzaldehyde
Formel: C₇H₄ClFO
Molecular Weight: 158.56 g/mol
MDL: MFCD01631571
CAS-Nummer: 85070-48-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR