3-Chlor-2-fluorbenzamid

Lieferant: FLUOROCHEM

Synonyme: 3-Chlor-2-fluorbenzoesäureamid

CFF922. 005615-5G 005615-10G
CHMPCFF922.EA 0 EUR
CHMPCFF922. FLUO005615-5G FLUO005615-10G
3-Chlor-2-fluorbenzamid
3-Chlor-2-fluorbenzamid
Formel: C₇H₅ClFNO
Molecular Weight: 173.57 g/mol
MDL: MFCD00042205
CAS-Nummer: 104326-94-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR