2-Chlor-4-fluorbenzylalkohol

Lieferant: FLUOROCHEM

005623-1G 005623-5G
FLUO005623-1GEA 30.5 EUR
FLUO005623-1G FLUO005623-5G
2-Chlor-4-fluorbenzylalkohol
2-Chlor-4-fluorbenzylalkohol
Formel: C₇H₆ClFO
Molecular Weight: 160.58 g/mol
MDL: MFCD00236024
CAS-Nummer: 208186-84-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR