3-Chlor-4-fluorzimtsäure
Lieferant: FLUOROCHEM
Synonyme:
3-(3-Chlor-4-fluorphenyl)propensäure, 3-Chlor-4-fluorcinnamsäure
005637-1G
005637-5G
FLUO005637-1GEA
39.9
EUR
FLUO005637-1G
FLUO005637-5G
3-Chlor-4-fluorzimtsäure
3-Chlor-4-fluorzimtsäure
Formel:
C₉H₆ClFO₂ Molecular Weight: 200.6 g/mol |
MDL:
MFCD01631393 CAS-Nummer: 155814-22-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...