2,4-Dichlor-5-fluorbenzamid

Lieferant: FLUOROCHEM

005728-5G 005728-1G
FLUO005728-5GEA 264 EUR
FLUO005728-5G FLUO005728-1G
2,4-Dichlor-5-fluorbenzamid
2,4-Dichlor-5-fluorbenzamid
Formel: C₇H₄Cl₂FNO
Molecular Weight: 208.02 g/mol
MDL: MFCD01631478
CAS-Nummer: 135748-33-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR