2,6-Difluor-3-methylanisol

Lieferant: FLUOROCHEM

005745-1G 005745-5G
FLUO005745-1GEA 67.1 EUR
FLUO005745-1G FLUO005745-5G
2,6-Difluor-3-methylanisol
2,6-Difluor-3-methylanisol
Formel: C₈H₈F₂O
Molecular Weight: 158.15 g/mol
MDL: MFCD01631341
CAS-Nummer: 261763-33-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR