4-Fluor-3-toluoylchlorid

Lieferant: FLUOROCHEM

Synonyme: 4-Fluor-3-tolylsäurechlorid, 4-Fluor-3-methylbenzoesäurechlorid, 4-Fluor-3-methylbenzol-1-carbonylchlorid, 4-Fluor-3-methylbenzoylchlorid

005792-5G 005792-1G
FLUO005792-5GEA 222 EUR
FLUO005792-5G FLUO005792-1G
4-Fluor-3-toluoylchlorid
4-Fluor-3-toluoylchlorid
Formel: C₈H₆ClFO
Molecular Weight: 172.59 g/mol
Siedepunkt: 110…115 °C (20 mmHg)
Lagertemperatur: Kühlschrank
MDL: MFCD01631426
CAS-Nummer: 455-84-5
UN: 3265
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR