3-Fluor-4-methylbenzylalkohol

Lieferant: FLUOROCHEM

Synonyme: (3-Fluoro-4-methylphenyl)methanol

005793-1G 005793-5G
FLUO005793-1GEA 0 EUR
FLUO005793-1G FLUO005793-5G
3-Fluor-4-methylbenzylalkohol
3-Fluor-4-methylbenzylalkohol
Formel: C₈H₉FO
MDL: MFCD01631533
CAS-Nummer: 192702-79-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR