3-Fluor-4-methylbenzylbromid
Lieferant: FLUOROCHEM
Synonyme:
4-(Bromomethyl)-2-fluoro-1-methylbenzene
005799-1G
005799-5G
FLUO005799-1GEA
77.1
EUR
FLUO005799-1G
FLUO005799-5G
3-Fluor-4-methylbenzylbromid
3-Fluor-4-methylbenzylbromid
Formel:
C₈H₈BrF Molecular Weight: 203.05 g/mol |
MDL:
MFCD01631534 CAS-Nummer: 145075-44-1 UN: 3265 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...