3-(Trifluormethyl)-p-anisidin

Lieferant: FLUOROCHEM

Synonyme: 5-Amino-2-methoxybenzotrifluorid, 3-(Trifluormethyl)-4-methoxyanilin, 4-Amino-2-(trifluormethyl)anisol, 3-(Trifluormethyl)-4-anisidin, 4-Methoxy-3-(trifluormethyl)anilin

005826-5G 005826-10G 005826-1G
FLUO005826-5GEA 51.8 EUR
FLUO005826-5G FLUO005826-10G FLUO005826-1G
3-(Trifluormethyl)-p-anisidin
3-(Trifluormethyl)-p-anisidin
Formel: C₈H₈F₃NO
Molecular Weight: 191.15 g/mol
Siedepunkt: 112 °C (18 mmHg)
Schmelzpunkt: 59…60 °C
MDL: MFCD00043460
CAS-Nummer: 393-15-7
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR