α-Brom-2,3,4,5-tetrafluortoluol
Lieferant: FLUOROCHEM
|
Gefahr
|
Synonyme:
2,3,4,5-Tetrafluorobenzyl bromide
005845-5G
FLUO005845-5GEA
441
EUR
FLUO005845-5G
α-Brom-2,3,4,5-tetrafluortoluol
α-Brom-2,3,4,5-tetrafluortoluol
Formel:
C₇H₃BrF₄ Molecular Weight: 243 g/mol Siedepunkt: 107…110 °C (1013 hPa) |
MDL:
MFCD01631453 CAS-Nummer: 53001-71-1 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...