3-(Trifluormethoxy)anisol
Lieferant: FLUOROCHEM
|
Achtung
|
005855-1G
005855-5G
005855.10G
FLUO005855-1GEA
45.9
EUR
FLUO005855-1G
FLUO005855-5G
FLUO005855.10G
3-(Trifluormethoxy)anisol
3-(Trifluormethoxy)anisol
Formel:
C₈H₇F₃O₂ |
MDL:
MFCD01631504 CAS-Nummer: 142738-94-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...