2-Chlor-3-nitro-5-thiophensulfonylchlorid

Lieferant: FLUOROCHEM

CFF258.2.5 CFF258.10 CFF258.25 005880-5G
CHMPCFF258.2.5EA 268 EUR
CHMPCFF258.2.5 CHMPCFF258.10 CHMPCFF258.25 FLUO005880-5G
2-Chlor-3-nitro-5-thiophensulfonylchlorid
2-Chlor-3-nitro-5-thiophensulfonylchlorid
Formel: C₄HCl₂NO₄S₂
MDL: MFCD00067978
CAS-Nummer: 58457-24-2
UN: 3261
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR