5-(Brommethyl)benzofurazan
Lieferant: FLUOROCHEM
Synonyme:
5-(Bromomethyl)benzo[c][1,2,5]oxadiazole, 5-(Bromomethyl)-2,1,3-benzoxadiazole
CFF516.2.5
CFF516.10
005888-2.5G
CHMPCFF516.2.5EA
481
EUR
CHMPCFF516.2.5
CHMPCFF516.10
FLUO005888-2.5G
5-(Brommethyl)benzofurazan
5-(Brommethyl)benzofurazan
Formel:
C₇H₅BrN₂O Molecular Weight: 213,0314 g/mol |
MDL:
MFCD00174369 CAS-Nummer: 32863-31-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...