2,1,3-Benzothiadiazol-4-sulfonylchlorid

Lieferant: FLUOROCHEM

Synonyme: Benzo[c][1,2,5]thiadiazole-4-sulphonyl chloride

005896-2.5G
FLUO005896-2.5GEA 0 EUR
FLUO005896-2.5G
2,1,3-Benzothiadiazol-4-sulfonylchlorid
2,1,3-Benzothiadiazol-4-sulfonylchlorid
Formel: C₆H₃ClN₂O₂S₂
Molecular Weight: 234,68 g/mol
MDL: MFCD00068049
CAS-Nummer: 73713-79-8
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR