1-(4-Fluorphenyl)-2-pyrrolidinon

Lieferant: FLUOROCHEM

Synonyme: 1-(4-Fluorophenyl)pyrrolidin-2-one

005898-1G 005898-5G
FLUO005898-1GEA 60.9 EUR
FLUO005898-1G FLUO005898-5G
1-(4-Fluorphenyl)-2-pyrrolidinon
1-(4-Fluorphenyl)-2-pyrrolidinon
Formel: C₁₀H₁₀FNO
Molecular Weight: 179.19 g/mol
MDL: MFCD00138504
CAS-Nummer: 54660-08-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR