Trimethylsilyl-2,2-difluor-2-(fluorsulfonyl)acetat

Lieferant: FLUOROCHEM

005917-5G 005917-25G
FLUO005917-5GEA 115 EUR
FLUO005917-5G FLUO005917-25G
Trimethylsilyl-2,2-difluor-2-(fluorsulfonyl)acetat
Trimethylsilyl-2,2-difluor-2-(fluorsulfonyl)acetat
Formel: C₅H₉F₃O₄SSi
MDL: MFCD02093343
CAS-Nummer: 120801-75-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR