2-[(2-Chlorbenzyl)oxy]benzaldehyd

Lieferant: FLUOROCHEM

005963-5G
FLUO005963-5GEA 0 EUR
FLUO005963-5G
2-[(2-Chlorbenzyl)oxy]benzaldehyd
2-[(2-Chlorbenzyl)oxy]benzaldehyd
Formel: C₁₄H₁₁ClO₂
Molecular Weight: 246,689 g/mol
MDL: MFCD00442935
CAS-Nummer: 53389-99-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR