2-[(3-Chlorbenzyl)oxy]benzaldehyd

Lieferant: FLUOROCHEM

005964-5G
FLUO005964-5GEA 2250 EUR
FLUO005964-5G
2-[(3-Chlorbenzyl)oxy]benzaldehyd
2-[(3-Chlorbenzyl)oxy]benzaldehyd
Formel: C₁₄H₁₁ClO₂
MDL: MFCD03093784
CAS-Nummer: 40359-59-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR