5-Amino-4-cyan-3-methyl-1-phenyl-1H-pyrazol

Lieferant: FLUOROCHEM

005982-1G
FLUO005982-1GEA 121 EUR
FLUO005982-1G
5-Amino-4-cyan-3-methyl-1-phenyl-1H-pyrazol
5-Amino-4-cyano-3-methyl-1-phenyl-1H-pyrazole
Formel: C₁₁H₁₀N₄
MDL: MFCD00128285
CAS-Nummer: 5346-56-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR