3,4'-Bis(trifluormethyl)benzhydrol

Lieferant: FLUOROCHEM

006006-1G
FLUO006006-1GEA 84.9 EUR
FLUO006006-1G
3,4'-Bis(trifluormethyl)benzhydrol
3,4'-Bis(trifluormethyl)benzhydrol
Formel: C₁₅H₁₀F₆O
Molecular Weight: 320,23 g/mol
MDL: MFCD00045091
CAS-Nummer: 203915-48-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR