2,4-Bis(trifluormethyl)benzonitril

Lieferant: FLUOROCHEM

006011-1G 006011-5G
FLUO006011-1GEA 121 EUR
FLUO006011-1G FLUO006011-5G
2,4-Bis(trifluormethyl)benzonitril
2,4-Bis(trifluormethyl)benzonitril
Formel: C₉H₃F₆N
MDL: MFCD00061225
CAS-Nummer: 177952-38-4
UN: 3276
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR