2,4-Bis(trifluormethyl)benzylalkohol

Lieferant: FLUOROCHEM

006020-1G 006020-5G 006020-10G
FLUO006020-1GEA 39.9 EUR
FLUO006020-1G FLUO006020-5G FLUO006020-10G
2,4-Bis(trifluormethyl)benzylalkohol
2,4-Bis(trifluormethyl)benzylalkohol
Formel: C₉H₆F₆O
MDL: MFCD00042491
CAS-Nummer: 143158-15-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR