3,5-Bis(trifluormethyl)benzylamin
Lieferant: FLUOROCHEM
006021-25G
006021-5G
FLUO006021-25GEA
72.9
EUR
FLUO006021-25G
FLUO006021-5G
3,5-Bis(trifluormethyl)benzylamin
3,5-Bis(trifluormethyl)benzylamin
Formel:
C₉H₇F₆N Siedepunkt: 82…84 °C (15 mmHg) Schmelzpunkt: 46…55 °C |
MDL:
MFCD00009909 CAS-Nummer: 85068-29-7 EINECS: 285-290-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...