1-Brom-2,4-bis(trifluormethyl)benzol

Lieferant: FLUOROCHEM

Synonyme: 2,4-Bis(trifluoromethyl)-1-bromobenzene

006025-5G 006025-25G
FLUO006025-5GEA 55.2 EUR
FLUO006025-5G FLUO006025-25G
1-Brom-2,4-bis(trifluormethyl)benzol
1-Brom-2,4-bis(trifluormethyl)benzol
Formel: C₈H₃BrF₆
MDL: MFCD00074904
CAS-Nummer: 327-75-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR