(±)-Ethyl-2-brompropionat

Lieferant: FLUOROCHEM

Synonyme: DL-Ethyl 2-bromopropionate, Ethyl 2-bromopropionate

006095-500G 006095-2.5KG
FLUO006095-500GEA 125 EUR
FLUO006095-500G FLUO006095-2.5KG
(±)-Ethyl-2-brompropionat
(±)-Ethyl-2-brompropionat
Formel: C₅H₉BrO₂
MDL: MFCD00000144
CAS-Nummer: 535-11-5
EINECS: 208-609-5
UN: 1992
ADR: 3,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR