Fluorinert™ FC-43
Lieferant: FLUOROCHEM
006162-1KG
FLUO006162-1KGEA
0
EUR
FLUO006162-1KG
Fluorinert™ FC-43
Fluorinert™
Bitte beachten Fluorinert is a trademark of 3M Company
Siedepunkt:
155 °C (1013 hPa) Schmelzpunkt: –55 °C Dichte: 1,883 g/cm³ (20 °C) |
MDL:
MFCD00000436 CAS-Nummer: 311-89-7 EINECS: 206-223-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...