2-Fluoracetanilid

Lieferant: FLUOROCHEM

006173-100G
FLUO006173-100GEA 107 EUR
FLUO006173-100G
2-Fluoracetanilid
2-Fluoracetanilid
Formel: C₈H₈FNO
Molecular Weight: 153.16
CAS-Nummer: 330-68-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR