4-Fluorbenzolsulfonamid

Lieferant: FLUOROCHEM

006184-5G 006184-25G 006184-1G
FLUO006184-5GEA 31.1 EUR
FLUO006184-5G FLUO006184-25G FLUO006184-1G
4-Fluorbenzolsulfonamid
4-Fluorbenzolsulfonamid
Formel: C₆H₆FNO₂S
MDL: MFCD00025384
CAS-Nummer: 402-46-0
EINECS: 206-946-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR