α-Brom-2-fluortoluol
Lieferant: FLUOROCHEM
006200-25G
006200-100G
006200-250G
FLUO006200-25GEA
30.8
EUR
FLUO006200-25G
FLUO006200-100G
FLUO006200-250G
α-Brom-2-fluortoluol
α-Brom-2-fluortoluol
Formel:
C₇H₆BrF Siedepunkt: 84 °C (1013 hPa) Dichte: 1,567 g/cm³ (20 °C) |
MDL:
MFCD00000324 CAS-Nummer: 446-48-0 EINECS: 207-169-1 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...