1-Fluor-2,6-dichlorpyridiniumtrifluormethansulfonat
Lieferant: FLUOROCHEM
Synonyme:
2,6-Dichloro-1-fluoropyridin-1-ium trifluoromethanesulphonate
006226-5G
FLUO006226-5GEA
0
EUR
FLUO006226-5G
1-Fluor-2,6-dichlorpyridiniumtrifluormethansulfonat
1-Fluor-2,6-dichlorpyridiniumtrifluormethansulfonat
Formel:
C₆H₃Cl₂F₄NO₃S Molecular Weight: 316,575 g/mol |
MDL:
MFCD00191438 CAS-Nummer: 130433-68-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...