1-Fluor-4-phenoxybenzol

Lieferant: FLUOROCHEM

006228-5G
FLUO006228-5GEA 72.9 EUR
FLUO006228-5G
1-Fluor-4-phenoxybenzol
1-Fluor-4-phenoxybenzol
Formel: C₁₂H₉FO
Molecular Weight: 188,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00055239
CAS-Nummer: 330-84-7
EINECS: 206-357-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR