1-Fluor-4-iod-3-nitrobenzol

Lieferant: FLUOROCHEM

Synonyme: 5-Fluor-2-iodnitrobenzol, 4-Fluor-1-iod-2-nitrobenzol

CFF1115. 006246-5G 006246-1G 006246-25G
CHMPCFF1115.EA 0 EUR
CHMPCFF1115. FLUO006246-5G FLUO006246-1G FLUO006246-25G
1-Fluor-4-iod-3-nitrobenzol
1-Fluoro-4-iodo-3-nitrobenzene
Formel: C₆H₃FINO₂
Molecular Weight: 267 g/mol
Siedepunkt: 162…163 °C (35 mmHg)
Schmelzpunkt: 22,5
MDL: MFCD00153169
CAS-Nummer: 364-77-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR